1,3-Bis(tosyloxy)propane ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:5469-66-9
:
Formula:C₁₇H₂₀O₆S₂
MDL Number:MFCD00013392
:384.46 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:90.0 to 93.0 deg-C
Specification Test Results
Frequently Bought Together