
2-[N,N-Bis(trifluoromethanesulphonyl)amino]-5-chloropyridine ≥96.0% (by GC) triflating reagent
Supplier: TCI
Certificates
About this item
CAS Number:145100-51-2
:
Formula:C₇H₃ClF₆N₂O₄S₂
MDL Number:MFCD00191833
:392.67 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together