O-Benzyl-N-(tert-butoxycarbonyl)-D-serine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:47173-80-8
:
Formula:C₁₅H₂₁NO₅
MDL Number:MFCD00038248
:295.34 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:58.0 to 62.0 deg-C
- Specific rotation [a]20/D:-18.0 to -21.0 deg (C=2, EtOH (80%))
Specification Test Results
Frequently Bought Together