
1,2-Bis(tert-butylthio)ethane borane ≥90.0% (by titrimetric analysis)
Catalog # TCIAB1827-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 90.0 %
Specification Test Results
About this item
:
:234.07 g/mol
EINECS:275-595-5
:GHS07
Storage Temperature:Refrigerator
CAS Number:71522-78-6
Formula:C₁₀H₂₈B₂S₂
MDL Number:MFCD00674193