Order Entry
Netherlands
ContactUsLinkComponent
Bis(trimethylsilyl)methylamine ≥95.0% (by GC)
Flammable
Corrosive
Bis(trimethylsilyl)methylamine ≥95.0% (by GC)
Catalog # TCIAB1826-1G
Supplier:  TCI
CAS Number:  
Flammable
Corrosive
Bis(trimethylsilyl)methylamine ≥95.0% (by GC)
Catalog # TCIAB1826-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :175.42 g/mol
Boiling Pt:70°C
UN:2920
ADR:8,II
 :GHS02
 :GHS05
CAS Number:134340-00-4
Formula:C₇H₂₁NSi₂
MDL Number:MFCD00008262