Order Entry
Netherlands
ContactUsLinkComponent
BMH (1,6-Bis(maleimido)hexane) ≥97.0% (by HPLC, total nitrogen)
 
BMH (1,6-Bis(maleimido)hexane) ≥97.0% (by HPLC, total nitrogen)
  TCIAB1787-100MG
 :  TCI
 :  
 
BMH (1,6-Bis(maleimido)hexane) ≥97.0% (by HPLC, total nitrogen)
  TCIAB1787-100MG
 :  TCI
 :  

 

  • Pk:
    100 mg
  • Seq:
    2440

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 96.0 %

 

 

 :
 :276.29 g/mol
 :225-449-1
 :GHS07
 :4856-87-5
 :C₁₄H₁₆N₂O₄
 :MFCD00047122