

N,N'-Bis(vinylsulphonylacetyl)ethylenediamine ≥95.0% (by total nitrogen basis)
Catalog # TCIAB1746-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 95.0 %
- Solubility in hot Methanol:almost transparency
Specification Test Results
About this item
:
:324.37 g/mol
EINECS:404-790-1
:GHS05
:GHS07
CAS Number:66710-66-5
Formula:C₁₀H₁₆N₂O₆S₂
MDL Number:MFCD00671499