O-tert-Butyl-N-carbobenzoxy-L-serine methyl ester ≥98.0% (by total nitrogen basis)
Catalog # TCIAB1732-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:41.0 to 45.0 deg-C
- Specific rotation [a]20/D:+3.0 to +8.0 deg (C=2, EtOH)
Specification Test Results
About this item
:
:309.36 g/mol
CAS Number:1872-59-9
Formula:C₁₆H₂₃NO₅
MDL Number:MFCD00191072