Order Entry
Netherlands
ContactUsLinkComponent
3,9-Bis(2-cyanoethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane ≥98.0% (by GC)
Irritant
3,9-Bis(2-cyanoethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane ≥98.0% (by GC)
Catalog # TCIAB1723-25G
Supplier:  TCI
CAS Number:  
Irritant
3,9-Bis(2-cyanoethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane ≥98.0% (by GC)
Catalog # TCIAB1723-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :266.30 g/mol
EINECS:221-294-9
 :GHS07
CAS Number:3058-04-6
Formula:C₁₃H₁₈N₂O₄
MDL Number:MFCD00191394