
N-(tert-Butyl)hydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:57497-39-9
:
Formula:C₄H₁₁NO
MDL Number:MFCD00012598
:125.60 g/mol
EINECS:260-771-6
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:182.0 to 188.0 deg-C
Specification Test Results
Frequently Bought Together