Order Entry
Netherlands
ContactUsLinkComponent
Bis(8-quinolinolato)copper(II) ≥95.0% (by titrimetric analysis)
Irritant
Health
Environment
Bis(8-quinolinolato)copper(II) ≥95.0% (by titrimetric analysis)
Catalog # TCIAB1677-25G
Supplier:  TCI
CAS Number:  
Irritant
Health
Environment
Bis(8-quinolinolato)copper(II) ≥95.0% (by titrimetric analysis)
Catalog # TCIAB1677-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Yellow to Brown to Dark green powder to crystal
  • Purity (Chelometric Titration):
    min. 95.0 %

Specification Test Results

About this item

 :
 :351.85 g/mol
EINECS:233-841-9
UN:3077
ADR:9,III
 :GHS07
 :GHS08
 :GHS09
CAS Number:10380-28-6
Formula:C₁₈H₁₂CuN₂O₂
MDL Number:MFCD00067392