
4,4'-Bis(4-aminophenoxy)biphenyl ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13080-85-8
:
Formula:C₂₄H₂₀N₂O₂
MDL Number:MFCD00039151
:368.44 g/mol
:GHS07
Specification Test Results
- Appearance:White to Gray to Red powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:196.0 to 200.0 deg-C
Specification Test Results
Frequently Bought Together