Order Entry
Netherlands
ContactUsLinkComponent
(R)-3-(Benzylthio)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥98.0% (by HPLC, titration analysis)
(R)-3-(Benzylthio)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB1666-5G
Supplier:  TCI
CAS Number:  
(R)-3-(Benzylthio)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB1666-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    86.0 to 88.0 deg-C
  • Specific rotation [a]20/D:
    -44.0 to -46.0 deg (C=1, AcOH)

Specification Test Results

About this item

 :
 :311.4 g/mol
EINECS:225-772-8
CAS Number:5068-28-0
Formula:C₁₅H₂₁NO₄S
MDL Number:MFCD00065567