Order Entry
Netherlands
ContactUsLinkComponent
N-(tert-Butoxycarbonyl)-L-methionine ≥98.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-L-methionine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB1636-10G
Supplier:  TCI
CAS Number:  
N-(tert-Butoxycarbonyl)-L-methionine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB1636-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    48.0 to 52.0 deg-C
  • Specific rotation [a]20/D:
    -20.0 to -24.0 deg (C=1.3, AcOH)

Specification Test Results

About this item

 :
 :249.33 g/mol
EINECS:219-639-3
CAS Number:2488-15-5
Formula:C₁₀H₁₉NO₄S
MDL Number:MFCD00065586