1,3-Bis(2-hydroxyethoxy)benzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:102-40-9
:
Formula:C₁₀H₁₄O₄
MDL Number:MFCD00016566
:198.22 g/mol
Boiling Pt:234°C
EINECS:203-028-3
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:90.0 to 95.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together