Order Entry
Netherlands
ContactUsLinkComponent
(+)-4,5-Bis[hydroxy(diphenyl)methyl]-2-methyl-2-phenyl-1,3-dioxolane ≥95.0% (by HPLC)
(+)-4,5-Bis[hydroxy(diphenyl)methyl]-2-methyl-2-phenyl-1,3-dioxolane ≥95.0% (by HPLC)
Catalogus # TCIAB1615-1G
Supplier:  TCI
CAS Number:  
undefined
(+)-4,5-Bis[hydroxy(diphenyl)methyl]-2-methyl-2-phenyl-1,3-dioxolane ≥95.0% (by HPLC)
Catalogus # TCIAB1615-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Melting point:
    118.0 to 122.0 deg-C
  • Specific rotation [a]20/D:
    +81.0 to +89.0 deg (C=1, CHCL3)

Specification Test Results

About this item

 :
 :528.65 g/mol
CAS Number:109306-21-0
Formula:C₃₆H₃₂O₄
MDL Number:MFCD00191611