
4-Cyanophenyl-4-(3-butenyloxy)benzoate ≥97.0% (by GC)
TCIAB1586-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:82.0 to 86.0 deg-C
- Solubility in hot Methanol:almost transparency
:
:293.32 g/mol
:GHS07
:114482-57-4
:C₁₈H₁₅NO₃
:MFCD00191384