
cis-1,2-Bis(phenylsulphonyl)ethylene ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:963-15-5
:
Formula:C₁₄H₁₂O₄S₂
MDL Number:MFCD00010140
:308.37 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
Specification Test Results
Frequently Bought Together