Order Entry
Netherlands
ContactUsLinkComponent
1,3-Bis(3-aminopropyl)tetramethyldisiloxane ≥83.0% (by GC) monomer for silicon modified polyamides
Corrosive
1,3-Bis(3-aminopropyl)tetramethyldisiloxane ≥83.0% (by GC) monomer for silicon modified polyamides
Supplier:  TCI
Certificates
Corrosive
1,3-Bis(3-aminopropyl)tetramethyldisiloxane ≥83.0% (by GC) monomer for silicon modified polyamides
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 83.0 %

Specification Test Results

Related Information

Frequently Bought Together