Order Entry
Netherlands
ContactUsLinkComponent
1,4-Bis(4-methylstyryl)benzene ≥98.0% (by HPLC)
1,4-Bis(4-methylstyryl)benzene ≥98.0% (by HPLC)
Catalog # TCIAB1550-5G
Supplier:  TCI
CAS Number:  
1,4-Bis(4-methylstyryl)benzene ≥98.0% (by HPLC)
Catalog # TCIAB1550-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%

Specification Test Results

About this item

 :
 :310.44 g/mol
CAS Number:76439-00-4
Formula:C₂₄H₂₂
MDL Number:MFCD00191317