
trans-1,2-Dicyano-1,2-bis(2,4,5-trimethyl-3-thienyl)ethene ≥97.0% (by total nitrogen basis)
TCIAB1537-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:166.0 to 170.0 deg-C
:
:326.48 g/mol
:3439
:6.1,III
:GHS06
:112440-50-3
:C₁₈H₁₈N₂S₂
:MFCD00145417