Ethyl-4-(butylamino)benzoate ≥98.0% (by GC)
TCIAB1472-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:69.0 to 72.0 deg-C
- Solubility in Methanol:very faint turbidity
:
:221.30 g/mol
:220°C
:202-322-9
:94-32-6
:C₁₃H₁₉NO₂
:MFCD00017283