1,3-Bis(4-methoxyphenoxy)benzene ≥98.0% (by GC)
Catalog # TCIAB1454-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:83.0 to 86.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
About this item
:
:322.36 g/mol
CAS Number:13118-91-7
Formula:C₂₀H₁₈O₄
MDL Number:MFCD00142506