

1,3-Bis(2-isocyanato-2-propyl)benzene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2778-42-9
:
Formula:C₁₄H₁₆N₂O₂
MDL Number:MFCD00142493
:244.29 g/mol
Boiling Pt:106°C
Melting Pt:–10°C
Flash Pt:153°C
Storage Temperature:Refrigerator
EINECS:220-474-4
UN:2810
ADR:6.1,I
:GHS06
:GHS09
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together