Order Entry
Netherlands
ContactUsLinkComponent
 
 
1,3-Bis(dicyanomethylidene)indan ≥98.0% (by HPLC, total nitrogen)
 :  TCI
 
undefined
 
1,3-Bis(dicyanomethylidene)indan ≥98.0% (by HPLC, total nitrogen)
 :  TCI

 

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

 

Frequently Bought Together