Order Entry
Netherlands
ContactUsLinkComponent
 
Bisabolene mixture
Catalog # TCIAB1413-25ML
Supplier:  TCI
CAS Number:  
Bisabolene mixture
Catalog # TCIAB1413-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Purity (GC):
    multi component mixture

Specification Test Results

About this item

 :
 :204.36 g/mol
CAS Number:495-62-5
Formula:C₁₅H₂₄
MDL Number:MFCD00129080