
4-tert-Butyl-3-methoxy-2,6-dinitrotoluene ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:83-66-9
:
Formula:C₁₂H₁₆N₂O₅
MDL Number:MFCD00024268
:268.27 g/mol
Boiling Pt:185°C
EINECS:201-493-7
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:85.0 to 87.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together