

Bis(tetrabutylammonium) bis(maleonitriledithiolato)nickel(II) complex ≥97.0% (by total nitrogen and titration analysis)
Catalog # TCIAB1371-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Orange to Red powder to crystal
- Purity (with Total Nitrogen):min. 97.0 %
- Purity (Chelometric Titration):min. 97.0 %
Specification Test Results
About this item
:
ADR:6.1,III
:823.99 g/mol
UN:3439
:GHS06
:GHS08
CAS Number:18958-57-1
Formula:C₄₀H₇₂N₆NiS₄
MDL Number:MFCD00059041