
4-(2-Benzothiazolyldithio)morpholine ≥95.0% (by total nitrogen basis)
Catalog # TCIAB1349-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Purity (with Total Nitrogen):min. 95.0 %
- Melting point:130.0 to 134.0 deg-C
Specification Test Results
About this item
:
:284.41 g/mol
EINECS:202-410-7
UN:3077
ADR:9,III
:GHS09
CAS Number:95-32-9
Formula:C₁₁H₁₂N₂OS₃
MDL Number:MFCD00059033