

2,2-Bis(4-chloroformyloxyphenyl)propane ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2024-88-6
:
Formula:C₁₇H₁₄Cl₂O₄
MDL Number:MFCD00060109
:353.20 g/mol
Boiling Pt:216°C
EINECS:217-970-8
UN:2923
ADR:8,III
:GHS05
:GHS06
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Argentmetric Titration):min. 97.0 %
- Melting point:93.0 to 96.0 deg-C
- Solubility in Acetone:almost transparency
Specification Test Results
Frequently Bought Together