
4,4'-Biphenyldicarbonyl chloride ≥97.0% (by GC, titration analysis)
Catalog # TCIAB1308-10G
Supplier: TCI
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystaline
- Purity (GC):min. 97.0 %
- Purity (Argentmetric Titration):min. 97.0 %
- Melting point:186.0 to 190.0 deg-C
- Solubility in hot Toluene:very faint turbidity
Specification Test Results
About this item
:
:279.12 g/mol
EINECS:219-085-2
UN:3261
ADR:8,II
:GHS05
CAS Number:2351-37-3
Formula:C₁₄H₈Cl₂O₂
MDL Number:MFCD00058934