
1,2-Bis(trichlorosilyl)ethane ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2504-64-5
:
Formula:C₂H₄Cl₆Si₂
MDL Number:MFCD00013601
:296.92 g/mol
EINECS:219-710-9
UN:2987
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (Argentmetric Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together