cis-4-tert-Butylcyclohexanol ≥98.0% (by GC)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:937-05-3
:
Formule:C₁₀H₂₀O
MDL-nummer:MFCD00070476
:156.27 g/mol
EINECS:213-321-8
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:80.0 to 84.0 deg-C
- Solubility in Methanol:almost transparency
Specificatie testresultaten
Frequently Bought Together