
Bis(3-aminopropyl)ether ≥98.0% (by GC, titration analysis)
Catalog # TCIAB1268-5ML
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
About this item
:
Boiling Pt:72°C
UN:3267
ADR:8,III
:132.21 g/mol
:GHS05
CAS Number:2157-24-6
Formula:C₆H₁₆N₂O
MDL Number:MFCD00059961