Order Entry
Netherlands
ContactUsLinkComponent
2-Dimethylaminoethyl benzoate ≥96.0% (by GC, titration analysis)
Irritant
2-Dimethylaminoethyl benzoate ≥96.0% (by GC, titration analysis)
Catalog # TCIAB1266-25G
Supplier:  TCI
CAS Number:  
Irritant
2-Dimethylaminoethyl benzoate ≥96.0% (by GC, titration analysis)
Catalog # TCIAB1266-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Red to Green clear liquid
  • Purity (GC):
    min. 96.0 %
  • Purity (Nonaqueous Titration):
    min. 96.0 %

Specification Test Results

About this item

 :
 :193.25 g/mol
Boiling Pt:159°C
EINECS:218-630-1
 :GHS07
CAS Number:2208-05-1
Formula:C₁₁H₁₅NO₂
MDL Number:MFCD00051067