Order Entry
Netherlands
ContactUsLinkComponent
2,2'-Bis(2-oxazoline) ≥97.0% (by total nitrogen basis)
2,2'-Bis(2-oxazoline) ≥97.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
2,2'-Bis(2-oxazoline) ≥97.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Melting point:
    209.0 to 215.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

Frequently Bought Together