Order Entry
Netherlands
ContactUsLinkComponent
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride ≥98.0% (by HPLC, titration analysis)
Irritant
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB1224-25G
Supplier:  TCI
CAS Number:  
Irritant
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB1224-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    142.0 to 145.0 deg-C
  • Solubility in Water:
    almost transparency

Specification Test Results

About this item

 :
 :269.79 g/mol
EINECS:224-947-6
 :GHS07
CAS Number:4568-71-2
Formula:C₁₃H₁₆ClNOS
MDL Number:MFCD00011959