Order Entry
Netherlands
ContactUsLinkComponent
 
Bis[(2S,3aR,4S,7aR)octahydro-7,8,8-trimethyl-4,7-methanobenzofuran-2-yl]ether ≥98.0% (by HPLC) for optical resolution
  TCIAB1220-100MG
 :  TCI
 :  
Bis[(2S,3aR,4S,7aR)octahydro-7,8,8-trimethyl-4,7-methanobenzofuran-2-yl]ether ≥98.0% (by HPLC) for optical resolution
  TCIAB1220-100MG
 :  TCI
 :  

 

  • Pk:
    100 mg
  • Seq:
    2440

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    149.0 to 153.0 deg-C
  • Specific rotation [a]20/D:
    -200.0 to -205.0 deg (C=2, THF)

 

 

 :
 :374.57 g/mol
 :108031-79-4
 :C₂₄H₃₈O₃
 :MFCD00151131