

1,4-Butanediolbis(3-aminopropyl)ether ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:7300-34-7
:
Formula:C₁₀H₂₄N₂O₂
MDL Number:MFCD00009802
:204.31 g/mol
Boiling Pt:136°C
Flash Pt:159°C
EINECS:230-745-9
UN:2735
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together