Order Entry
Netherlands
ContactUsLinkComponent
 
1,1-Bis(morpholino)ethylene ≥97.0% (by GC, titration analysis)
Catalog # TCIAB1180-10G
Supplier:  TCI
CAS Number:  
1,1-Bis(morpholino)ethylene ≥97.0% (by GC, titration analysis)
Catalog # TCIAB1180-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to lump
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %

Specification Test Results

About this item

 :
 :198.27 g/mol
CAS Number:14212-87-4
Formula:C₁₀H₁₈N₂O₂
MDL Number:MFCD00059775