Order Entry
Netherlands
ContactUsLinkComponent
 
 
4-Methyl-2-phenyl-1,3-dioxolane (mixture of isomers) ≥98.0% (by GC)
  TCIAB1094-25ML
 :  TCI
 :  
 
4-Methyl-2-phenyl-1,3-dioxolane (mixture of isomers) ≥98.0% (by GC)
  TCIAB1094-25ML
 :  TCI
 :  

 

  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomer)

 

 

 :
 :164.20 g/mol
 :125°C
 :113°C
 :219-906-4
 :GHS07
 :2568-25-4
 :C₁₀H₁₂O₂
 :MFCD00059732