Order Entry
Netherlands
ContactUsLinkComponent
 
 
trans-2-Hexenyl butyrate ≥93.0% (by GC)
  TCIAB1075-25ML
 :  TCI
 :  
 
trans-2-Hexenyl butyrate ≥93.0% (by GC)
  TCIAB1075-25ML
 :  TCI
 :  

 

  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 93.0 %

 

 

 :
 :170.25 g/mol
 :86°C
 :258-515-3
 :GHS07
 :53398-83-7
 :C₁₀H₁₈O₂
 :MFCD00036546