Order Entry
Netherlands
ContactUsLinkComponent
1,4-Butanediolbis(3-aminocrotonate) ≥96.0% (by GC)
Irritant
1,4-Butanediolbis(3-aminocrotonate) ≥96.0% (by GC)
Supplier:  TCI
Certificates
Irritant
1,4-Butanediolbis(3-aminocrotonate) ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Nonaqueous Titration):
    min. 96.0 %
  • Melting point:
    147.0 to 151.0 deg-C
  • Solubility in N,N-DMF:
    very faint turbidity

Specification Test Results

Frequently Bought Together