Order Entry
Netherlands
ContactUsLinkComponent
cis-3-Hexen-1-ylbenzoate ≥98.0% (by GC)
Irritant
cis-3-Hexen-1-ylbenzoate ≥98.0% (by GC)
Catalog # TCIAB1039-25ML
Supplier:  TCI
CAS Number:  
Irritant
cis-3-Hexen-1-ylbenzoate ≥98.0% (by GC)
Catalog # TCIAB1039-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :204.27 g/mol
Boiling Pt:135°C
Flash Pt:147°C
EINECS:246-669-4
 :GHS07
CAS Number:25152-85-6
Formula:C₁₃H₁₆O₂
MDL Number:MFCD00036526