1,4-Bis(2-methylstyryl)benzene ≥99.0% (by GC) solute for liquid scintillation counting
Supplier: TCI
Certificates
About this item
CAS Number:13280-61-0
:
Formula:C₂₄H₂₂
MDL Number:MFCD00008529
:310.44 g/mol
EINECS:236-285-5
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:178.0 to 181.0 deg-C
Specification Test Results
Frequently Bought Together