
1,3-Bis(chloromethyl)tetramethyldisilazane ≥95.0% (by GC)
Catalog # TCIAB0990-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid to slightly cloudy liquid
- Purity (GC):min. 95.0 %
Specification Test Results
About this item
:
Boiling Pt:72°C
Flash Pt:88°C
UN:1760
ADR:8,III
:230.28 g/mol
EINECS:238-622-1
:GHS05
CAS Number:14579-91-0
Formula:C₆H₁₇Cl₂NSi₂
MDL Number:MFCD00000876