
3,3'-Dichloro-N,N'-bis(4-methoxybenzylidene)benzidine ≥98.0% (by titrimetric analysis)
TCIAB0878-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Chloroform:almost transparency
:
:489.40 g/mol
:GHS07
:3540-78-1
:C₂₈H₂₂Cl₂N₂O₂
:MFCD00059294