Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
1,3-Bis[1-(2-hydroxyethyl)-4-piperidyl]propane ≥98.0% (by GC)
Catalog # TCIAB0851-25G
Supplier:  TCI
CAS Number:  
Irritant
1,3-Bis[1-(2-hydroxyethyl)-4-piperidyl]propane ≥98.0% (by GC)
Catalog # TCIAB0851-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    93.0 to 97.0 deg-C
  • Solubility in Methanol:
    very faint turbidity

Specification Test Results

About this item

 :
Boiling Pt:219°C
 :298.47 g/mol
EINECS:241-981-7
 :GHS07
CAS Number:18073-84-2
Formula:C₁₇H₃₄N₂O₂
MDL Number:MFCD00023767