

Borneol ≥70% (by GC) (contains ca. 20% Isoborneol)
Supplier: TCI
Certificates
About this item
CAS Number:507-70-0
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00066426
:154.25 g/mol
Merck Index:14,01338
EINECS:208-080-0
UN:1312
ADR:4.1,III
:GHS02
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 70.0 %
- Melting point:198.0 to 208.0 deg-C
Specification Test Results
Frequently Bought Together