
Berberine sulphate hydrate ≥98.0% (by HPLC, titration analysis)
: TCI
:316-41-6
:
:C₄₀H₃₆N₂O₁₂S
:MFCD00031743
:768.79 g/mol
:14,01154
:206-258-2
:1544
:6.1,III
:GHS06
- Appearance:Light yellow to Yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:9.5 to 16.0 %
Frequently Bought Together